Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips

Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants.

Manufacturer: Thermo Scientific™ 2160PHR

 View more versions of this product

Catalog No. 13-811-138

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Length (English) 3.6in
    Length (Metric) 9.09cm
    For Use With (Equipment) Universal Fit
    Volume (Metric) 200μL
    Product Line ART
    Filtered Yes
    Model ART Barrier
    Description Hinged rack, Barrier tip, extended length
    Packaging 96 Tips/Tray, 8 Trays/Pack, 5 Packs
    Clean Claims Free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
    Sterility Sterile
    Color Natural

    Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.

    Features:

    • Stackable hinged racks save space in your lab
    • Sterile tips are certified free from RNase, DNase and pyrogen

    Recommended for:

    • Liquid Handling
    • Molecular Biology
    • PCR
    • Biotechnology

    • Purchase two cases of Thermo Scientific™ ART™ Tips in Hinged Racks and get an additional case at no cost. To take advantage of this offer, apply for the NLSU program.

      New Offer Buy Now

      Expires 2018-06-30 Redemption Only!

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit