Learn More
ART™ Barrier Hinged Rack Pipette Tips PROMO
Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants.
Manufacturer:  Thermo Scientific PR214005HR
 View more versions of this product
Catalog No. 02-682-259PR
  Check Availability
Add to cart
Description
Description
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Specifications
Universal Fit Pipettor
MicroPoint, extended length, reload insert, low retention
Filtered
ART 10 REACH
4.33cm
Universal Fit Pipettor
Hinged Rack
10μL
Sterile
1.71 in.
DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
ART