Learn More
Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips PROMO
Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants.
Manufacturer:  Thermo Scientific™ PR2149PHR
 View more versions of this product
Catalog No. 02-682-260PR
  Check Availability
Add to cart
Description
Description
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Specifications
Universal Fit Pipettor
MicroPoint, hinged racked, Barrier tip
Filtered
ART 20P
5.07cm
Universal Fit Pipettor
Hinged Rack
20μL
Sterile
2 in.
DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
ART