Learn More
Thermo Scientific™ ART™ Non-Filtered Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, which is designed for the way you work.
Manufacturer:  Thermo Scientific™ 3501HR
 View more versions of this product
Catalog No. 13-811-103
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Access tips with one hand using Thermo Scientific? ART? Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Description & Specifications
Specifications
ART Non-Filtered
No
Natural
96 Tips/Tray, 10 Trays/Pack, 5 Packs
3.13cm
Universal Fit
10μL
Sterile
ART Non-filtered
1.2in
RNase, DNase, ATP, and Pyrogens free
ART
Promotions
Promotions
  • Thermo Scientific
    Purchase two cases of Thermo Scientific™ ART™ Tips in Hinged Racks and get an additional case at no cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018
  • Thermo Scientific
    uy two cases of Thermo Scientific™ ART™ Barrier or Sterile Non-Filter Pipette Tips and get one case of equal or lesser value at no additional cost.
    06-30-2017