Thermo Scientific™ ART™ Non-Filtered Hinged Rack Pipette Tips

Access tips one-handed with this versatile hinged rack, which is designed for the way you work.

Manufacturer: Thermo Scientific™ 3771HR

 View more versions of this product

Catalog No. 13-811-145

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Length (English) 2.3in,2.3in
    Length (Metric) 5.84cm
    For Use With (Equipment) Universal Fit
    Volume (Metric) 300μL
    Product Line ART
    Filtered No
    Model ART Non-filtered
    Packaging 96 Tips/Tray, 10 Trays/Pack, 5 Packs,96 Tips/Tray, 10 Trays/Pack, 5 Packs
    Clean Claims RNase, DNase, ATP, and Pyrogens free
    Color Natural,Natural
    Description ART Non-Filtered
    Sterility Sterile
    Pipette Tip Brand ART Non-filtered
    Pipette Brand Universal
    Compatibility Universal

    Access tips with one hand using Thermo Scientific™ ART™ Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.

    Features:

    • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
    • Racks offer quick and easy access to pipette tips, saving valuable time
    • Front latch closes and locks, ensuring safe and secure storage throughout

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit