Learn More
Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
GREENER_CHOICE
Access tips one-handed with this versatile hinged rack, designed for the way you work.
$147.40 - $255.00
Specifications
Filtered Non-filtered
Color Natural
Model ART Non-filtered
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
View More Specs
Products 15
Catalog Number Mfr. No. Description Volume (Metric) Sterility Fit for Purpose Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) Sterility Fit for Purpose Price Quantity & Availability  
13-811-110
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3512HR
ART Non-filtered, Non-Filtered, Non-sterile, Rack 10 μL Non-sterile Certified Quality
Pack of 960 for $147.40
Case of 5 PK for $645.00
Add to Cart
13-811-112
GREENER_CHOICE
View Documents
Thermo Scientific™
351205HR
ART Low Retention, Non-Filtered, Non-sterile, Rack 10 μL Non-sterile Certified Quality
Pack of 960 for $164.40
Case of 5 PK for $718.00
Add to Cart
13-811-113
GREENER_CHOICE
View Documents
Thermo Scientific™
351105HR
ART Low Retention, Non-Filtered, Sterile, Rack 10 μL Sterile Sterile
Pack of 960 for $182.50
Case of 5 PK for $823.00
Add to Cart
13-811-134
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3542HR
ART Non-Filtered, Non-sterile, Rack 200 μL Non-sterile Certified Quality
Pack of 768 for $192.50
Case of 5 PK for $850.50
Edge
Add to Cart
13-811-135
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3541HR
ART Non-Filtered, Sterile, Rack 200 μL Sterile Sterile
Pack of 768 for $218.00
Case of 5 PK for $978.50
Edge
Add to Cart
13-811-136
GREENER_CHOICE
View Documents
Thermo Scientific™
354205HR
ART Low Retention, Non-Filtered, Non-sterile, Rack 200 μL Non-sterile Certified Quality
Pack of 768 for $176.60
Case of 5 PK for $796.00
Add to Cart
13-811-137
GREENER_CHOICE
View Documents
Thermo Scientific™
354105HR
ART Low Retention, Non-Filtered, Sterile, Rack 200 μL Sterile Sterile
Pack of 768 for $255.00
Case of 5 PK for $1,148.00
Edge
Add to Cart
13-811-166
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3792HR
ART Non-Filtered, Non-sterile, Rack 1000 μL Non-sterile Certified Quality
Pack of 768 for $154.90
Case of 4 PK for $562.50
Add to Cart
13-811-168
GREENER_CHOICE
View Documents
Thermo Scientific™
379205HR
ART Low Retention, Non-Filtered, Non-sterile, Rack 1000 μL Non-sterile Certified Quality
Pack of 768 for $164.20
Case of 4 PK for $597.00
Add to Cart
13-811-167
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3791HR
ART Non-Filtered, Sterile, Rack 1000 μL Sterile Sterile
Pack of 768 for $177.10
Case of 4 PK for $644.00
Add to Cart
13-811-169
GREENER_CHOICE
View Documents
Thermo Scientific™
379105HR
ART Low Retention, Non-Filtered, Sterile, Rack 1000 μL Sterile Sterile
Pack of 768 for $194.70
Case of 4 PK for $708.00
Add to Cart
13-811-172
GREENER_CHOICE
View Documents
Thermo Scientific™
3152HR
ART Non-Filtered, Non-sterile, Rack 1250 μL Non-sterile Certified Quality
Pack of 768 for $171.90
Case of 4 PK for $606.00
Add to Cart
13-811-173
GREENER_CHOICE
View Documents
Thermo Scientific™
3151HR
ART Non-Filtered, Sterile, Rack 1250 μL Sterile Sterile
Pack of 768 for $179.40
Case of 4 PK for $628.00
Add to Cart
13-811-174
GREENER_CHOICE
View Documents
Thermo Scientific™
315205HR
ART Low Retention, Non-Filtered, Non-sterile, Rack 1250 μL Non-sterile Certified Quality
Pack of 768 for $152.30
Case of 4 PK for $553.50
Add to Cart
13-811-175
GREENER_CHOICE
View Documents
Thermo Scientific™
315105HR
ART Low Retention, Non-Filtered, Sterile, Rack 1250 μL Sterile Sterile
Pack of 768 for $204.50
Case of 4 PK for $741.50
Add to Cart
Description
Description
Access tips with one hand using Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or the floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover — now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
Specifications
Non-filtered
ART Non-filtered
Rack
Universal Fit
Natural
DNase, RNase, DNA, ATP and Endotoxin Free
Universal Fit
ART
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.