Learn More
Thermo Scientific™ ART™ Non-Filtered Low Retention Universal Pipette Tips
Specifically designed for eliminating sample retention within the pipette tip
$205.00 - $213.00
Specifications
Sterility Sterile
Format Rack
Tip Style Low Retention
View More Specs
Products 2
Catalog Number Mfr. No. Format Volume (Metric) Tip Style Price Quantity & Availability  
Catalog Number Mfr. No. Format Volume (Metric) Tip Style Price Quantity & Availability  
21-403-519
View Documents
Thermo Scientific™
394105
Rack 0.5 to 200 μL Low Retention
Pack of 960 for $213.00
Case of 5 PK for $988.00
Add to cart
21-403-600
View Documents
Thermo Scientific™
358105
Rack 100 to 1000 μL Low Retention
Pack of 800 for $205.00
Case of 4 PK for $758.00
Edge
Add to cart
Description
Description
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting desired volume of liquid
  • Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
  • Other Thermo Scientific racked and bulk tips are nonsterile and autoclavable
Specifications
Specifications
Sterile
Rack
Low Retention