Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HydroLogix HLT Pipette Tips
A variety of tip styles and configurations
Supplier:  Thermo Scientific™ 395111
 View more versions of this product
Catalog No. 21-403-489
Add to Cart
Description
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipette tips onto the pipette. Accordingly, the forces required to eject the tips from your pipette are significantly reduced.
  • Available in blue or clear color
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
DNase, RNase, DNA, ATP and Endotoxin Free
ART
Non-Filtered
Blue
Rack
Product Suggestions
Product Suggestions