Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HydroLogix HLT Pipette Tips
A variety of tip styles and configurations
$75.70 - $146.40
Products 2
Catalog Number Mfr. No. Color Sterility Tip Style Format Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility Tip Style Format Price Quantity & Availability  
21-403-491
View Documents
Thermo Scientific™
395011
Blue Non-sterile Bulk
Pack of 1000 for $75.70
Case of 10 PK for $673.00
Add to cart
21-403-489
View Documents
Thermo Scientific™
395111
Blue Sterile Rack
Pack of 1000 for $146.40
Case of 4 PK for $543.50
Add to cart
Description
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.