Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HLT 5000uL Pipette Tips
A variety of tip styles and configurations
Manufacturer:  Thermo Scientific™ 3896
 View more versions of this product
Catalog No. 21-403-513
Add to cart
Description
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
Specifications
Specifications
5000 μL
Clear
Blunt
Non-sterile
Bulk