Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips, HLT Eppendorf-Style Tips
Style and packaging options
Manufacturer:  Thermo Scientific™ 3942
 View more versions of this product
Catalog No. 21-403-462
Add to cart
Description
Description
Style and packaging options
  • All sterile HLT branded tips are certified “Pure”
  • All nonsterile tips are manufactured in clean room environments and are certified DNA, RNase and DNase free
  • Stringent manufacturing and QA/QC testing procedures assure the most aseptic products on the market
  • Some tips have reference marks which provide a quick and convenient guide to visually establish whether you are pipeting the desired volume of liquid
Specifications
Specifications
200 μL
Yellow
Standard
Non-sterile
Rack