Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips, HLT Eppendorf-Style Tips
Style and packaging options
$93.50
Specifications
Volume (Metric) 200 μL
View More Specs
Products 1
Catalog Number Mfr. No. Color Sterility Tip Style Format Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility Tip Style Format Price Quantity & Availability  
21-403-462
View Documents
Thermo Scientific™
3942
Yellow Non-sterile Standard Rack Pack of 960 for $93.50
Case of 5 PK for $409.50
Add to cart
Description
Description
Style and packaging options
  • All sterile HLT branded tips are certified “Pure”
  • All nonsterile tips are manufactured in clean room environments and are certified DNA, RNase and DNase free
  • Stringent manufacturing and QA/QC testing procedures assure the most aseptic products on the market
  • Some tips have reference marks which provide a quick and convenient guide to visually establish whether you are pipeting the desired volume of liquid
Specifications
Specifications
200 μL