Learn More
Thermo Scientific™ ART™ Non-filtered Pipette Tips in Lift-off Lid Rack
Thermo Scientific™ ART™ non-filtered pipette tips, retain the high quality you expect from ART™ tips, but for applications where contamination is not a concern.
$137.50 - $169.60
Specifications
Autoclavable Autoclavable
Color Clear
Compatibility Universal Tip - Friction Fit, For use with most Pipette brands
Filtered Non-filtered
Disposable Yes - single use
View More Specs
Products 3
Catalog Number Mfr. No. Description Volume (Metric) Tip Style Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) Tip Style Price Quantity & Availability  
21-402-41
View Documents
Thermo Scientific™
3522
ART 20, Non-filtered, Non-sterile, Racked, Lift-off Lid 20 μL MicroPoint
Pack of 960 for $150.70
Case of 5 PK for $651.00
Add to cart
21-236-54
View Documents
Thermo Scientific™
3551
ART 200, Non-filtered, Sterile, Racked, Lift-off Lid 200 μL Universal Fit
Pack of 960 for $169.60
Case of 5 PK for $755.50
Add to cart
21-402-47
View Documents
Thermo Scientific™
3582
ART 1000 SoftFit, Non-filtered, Non-sterile, Racked, Lift-off Lid 1000 μL Universal Fit
Pack of 800 for $137.50
Case of 4 PK for $461.00
Add to cart
Description
Description
ART™ non-filtered tips deliver the same proven quality, performance and reliability, you have come to expect from ART™ barrier tips, but are designed for non-critical applications where contamination is not a concern. ART™ tips are a Universal Fit with friction-fit design and fit most pipette brands.
ART™ non-filtered tips deliver the same proven quality, performance and reliability you have come to expect from ART™ barrier tips
  • Non-filtered tips
    • ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
    • Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
Specifications
Autoclavable
Universal Tip - Friction Fit, For use with most Pipette brands
Yes - single use
Virgin Polypropylene
Clear
Non-filtered
Most brands of pipettes
ART