Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile

Click to view available options
Volume (Metric):
10 μL
1000 μL
20 μL
200 μL
Format:
Hinged Rack
Rack
Tip Style:
Low Retention
Standard
Fit for Purpose:
Certified Quality
Sterile
Description
Access tips with one hand using Thermo Scientific™ ART™ Non-filtered Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover - now you have a traditional lift-off when you need it.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
- Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
- Racks offer quick and easy access to pipette tips, saving valuable time
- Front latch closes and locks, ensuring safe and secure storage throughout

Specifications
Specifications
Volume (Metric) | 10 μL |
Filtered | Non-filtered |
Sterility | Sterile |
Color | Natural |
Autoclavable | Not Autoclavable |
Model | ART 10 REACH |
Length (English) | 1.7 in. |
Length (Metric) | 4.34 cm |
Clean Claims | DNase, RNase, DNA, ATP and Endotoxin Free |
Format | Rack |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction