Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile

Description
Access tips with one hand using Thermo Scientific™ ART™ Non-filtered Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover - now you have a traditional lift-off when you need it.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
- Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
- Racks offer quick and easy access to pipette tips, saving valuable time
- Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Not Autoclavable |
| Model | ART 200 |
| Length (English) | 2 in. |
| Length (Metric) | 5.07 cm |
| Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
| Format | Hinged Rack |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.