Learn More
Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks, non sterile PROMO
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Hinged Rack.
Manufacturer:  Thermo Scientific™ PR2006ER
 View more versions of this product
Catalog No. 02-682-271PR
  Check Availability
Add to cart
Description
Description
Features:
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
  • Certifications:
  • Nonsterile products certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Sterile products certified to be free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
  • Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
  • Specifications
    Specifications
    Certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
    Hinged Rack, 10 trays/pack, 5 packs/case
    Empty Hinged Racks
    Medium
    Non-sterile