Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART2.2/Art2b Antibody (Nika102), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065853B
Description
ART2.2/Art2b Monoclonal antibody specifically detects ART2.2/Art2b in Mouse samples. It is validated for Flow CytometrySpecifications
ART2.2/Art2b | |
Monoclonal | |
Biotin | |
ADP-ribosyltransferase 2b, Rt6, Rt-6, Rt6.2, Rt6-2 | |
pME.CD8LF-ART2.2 | |
0.125 mL | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
Nika102 | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
11872 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction