Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | Artemin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
Artemin Polyclonal specifically detects Artemin in Human samples. It is validated for Western Blot.Specifications
Artemin | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
PBS with 0.02% Sodium Azide | |
artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
ARTN | |
IgG | |
Affinity Purified | |
Reacts with the internal sequence of human Artemin. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q5T4W7 | |
9048 | |
Amino acids GPGSRARAAGARGC of human Artemin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
18 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title