Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemin Rat anti-Mouse, DyLight 405, Clone: RM0074-6G32, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121701V
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Artemin | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARTN | |
Mouse recombinant Artemin | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Artemin. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
RM0074-6G32 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
Rat | |
Protein A or G purified | |
Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
9048 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title