Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemisinin, Tocris Bioscience™
Click to view available options
Quantity:
50mg
Description
Antimalarial agent; interacts with heme to produce carbon-centred free radicals, causes protein alkylation and damages parasite microorganelles and membranes. Also selectively inhibits the P-type ATPase (PfATP6) of Plasmodium falciparum (Ki ∽ 150 nM). Displays antiangiogenic effects in mouse embryonic stem cell-derived embryoid bodies.
Specifications
Quantity | 50mg |
Formula Weight | 282.33 |
Percent Purity | >99% |
Chemical Name or Material | Artemisinin |
Safety and Handling
Recommended Storage : Store at +4°C