Learn More
Abnova™ ARTN (Human) ELISA Kit
For the Quantitative detection of Human ARTN.
Manufacturer:  Abnova™ KA4781
Catalog No. 89-935-769
Add to cart
Description
Description
  • ARTN (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human ARTN.
Specifications
Specifications
Human
artemin
Store the kit at 4°C. Avoid repeated freezing and thawing.
1 Kit
Kit
9048
Human
ARTN