Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARTS Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc SEPT401AP

Description
ARTS Polyclonal antibody specifically detects ARTS in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
ARTS | |
Polyclonal | |
Unconjugated | |
SEPTIN4 | |
4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47 | |
Synthetic peptide taken within amino acid region 400-450 on SEPT4 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O43236, P28661 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18952, 287606, 5414 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction