Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARX Goat anti-Human, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP275093

Catalog No. NBP275093


Add to cart

Description

Description

ARX Polyclonal antibody specifically detects ARX in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ARX
0.5 mg/ml
Western Blot 1:100-1:2000, Peptide ELISA
Purified
ARX
Peptide with sequence C-QVSISRSKSYREN, from the internal region of the protein sequence according to NP_620689.1.
58.2 kDa
100 μg
Primary
170302.0
ELISA, Western Blot
Unconjugated
TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
aristaless related homeobox, Aristaless-related homeobox, aristaless-related homeobox, X-linked, cancer/testis antigen 121, CT121, EIEE1, homeobox protein ARX, ISSXmental retardation, X-linked 36, mental retardation, X-linked 29, mental retardation, X-linked 32, mental retardation, X-linked 33, mental retardation, X-linked 38, mental retardation, X-linked 43, mental retardation, X-linked 54, mental retardation, X-linked 76, mental retardation, X-linked 87, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
Goat
IgG
Antigen Affinity-purified
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit