Learn More
Enzo Life Sciences Aryl hydrocarbon receptor polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2100100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant protein corresponding to aa 1-402 of the N-terminal B-1 AhR (Aryl hydrocarbon receptor). UniProt: P30561. Source: Purified from rabbit serum. Species Reactivity: Mouse. Crossreactivity: Does not cross-react with Arnt. Applications: IP, WB, ChIP. Recommended dilutions: Western Blot (1:2,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In TBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC0147433