Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase G/ARSG Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Arylsulfatase G/ARSG |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179576
|
Novus Biologicals
NBP179576 |
100 μL |
Each of 1 for $436.00
|
|
Description
Arylsulfatase G/ARSG Polyclonal specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Western Blot.Specifications
Arylsulfatase G/ARSG | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
arylsulfatase G, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG | |
ARSG | |
IgG | |
Affinity Purified | |
58 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_082986 | |
22901 | |
The immunogen for this antibody is Arsg. Peptide sequence GRDVSEVLFGKSQMGHRVLFHPNSGAAGEYGALQTVRLNHYKAFYITGGA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title