Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AS 1892802, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
928320-12-1
C20H19N3O2
333.391
WDTFYYZHMRBVHK-LJQANCHMSA-N
1-1s-2-hydroxy-1-phenylethyl-3-4-4-pyridinyl phenyl urea,3-1s-2-hydroxy-1-phenylethyl-1-4-pyridin-4-yl phenyl urea,n-1s-2-hydroxy-1-phenylethyl-n'-4-4-pyridinyl phenyl-urea
46911016
1-[(1S)-2-hydroxy-1-phenylethyl]-3-(4-pyridin-4-ylphenyl)urea
C1=CC=C(C=C1)C(CO)NC(=O)NC2=CC=C(C=C2)C3=CC=NC=C3
This item is not returnable. View return policy
928320-12-1
C20H19N3O2
333.391
WDTFYYZHMRBVHK-LJQANCHMSA-N
1-1s-2-hydroxy-1-phenylethyl-3-4-4-pyridinyl phenyl urea,3-1s-2-hydroxy-1-phenylethyl-1-4-pyridin-4-yl phenyl urea,n-1s-2-hydroxy-1-phenylethyl-n'-4-4-pyridinyl phenyl-urea
46911016
1-[(1S)-2-hydroxy-1-phenylethyl]-3-(4-pyridin-4-ylphenyl)urea
C1=CC=C(C=C1)C(CO)NC(=O)NC2=CC=C(C=C2)C3=CC=NC=C3

Potent ROCK inhibitor; orally bioavailable

Potent, ATP-competitive ROCK inhibitor (IC50 values are 52, 57 and 122 nM for human ROCK2, rat ROCK2 and human ROCK1 respectively by ELISA); also inhibits PKAC-α and PRKX (IC50 values are 200 and 325 nM respectively). Exhibits analgesic effects in rat models of inflammatory (AIA) and noninflammatory (MIA) arthritic pain. Orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 333.38
Percent Purity >99%
Chemical Name or Material AS 1892802

Recommended Storage : Store at Room Temperature