Tocris™ AS 1949490

SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) inhibitor

Manufacturer: Tocris™ 3718/10

 View more versions of this product

Catalog No. 37-181-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors AS 1949490
    Purity >99
    Molecular Weight 371.88
    Quantity 10mg
    Product Type AS 1949490

    Selective SHIP2 (SH2 domain-containing inositol 5'-phosphatase 2) inhibitor (IC50 values are 0.34µM and 0.62µM for mouse and human respectively); displays approximately 30-fold affinity for SHIP2 over SHIP1. Increases insulin-induced phosphorylation of Akt in L6 myotubules. Stimulates activation of glucose metabolism; regulates gluconeogenesis in vitro and in vivo and exhibits antidiabetic effects.

    • Chemical Name: 3-[(4-Chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide
    • Formula: C20H18ClNO2S
    • CAS Number: 1203680-76-5
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit