Tocris™ AS 2034178

Selective FFA1 (GPR40) agonist

Manufacturer: Tocris™ 5035/50

 View more versions of this product

Catalog No. 50-355-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors AS 2034178
    Purity >98
    Molecular Weight 448.53
    Quantity 50mg
    Product Type AS 2034178

    Selective free fatty acid receptor 1 (FFA1/GPR40) agonist. Exhibits selectivity for GPR40 over GPR41, GPR43, GPR119, GPR120 and PPARγ. Induces glucose-dependent insulin secretion in pancreatic MIN6 cells and in vivo. Enhances insulin sensitivity in a type 2 diabetes model in vivo. Orally available.

    • Chemical Name: 2-Fluoro-4-[[[1,2,3,4-Tetrahydro-1-(2-phenoxyethyl)-5-quinolinyl]methyl]amino]benzenepropanoic acid
    • Formula: C27H29FN2O3
    • CAS Number: 1030846-42-4
    • Solubility: Soluble to 50mM in DMSO and to 20mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit