Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ AS 605240

Potent and selective PI 3-kinase gamma (PI 3-Kgamma) inhibitor

Manufacturer:  Tocris™ 3578/50

 View more versions of this product

Catalog No. 35-785-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent and selective inhibitor of PI 3-kinase γ (PI 3-Kγ) (IC50 = 8nM). Displays 30-fold selectivity over PI 3-Kδ and PI 3-Kβ and 7.5-fold selectivity over PI 3-Kα. Suppresses the progression of joint inflammation and damage in both lymphocyte-independent and lymphocyte-dependent mouse models of rheumatoid arthritis. Orally active and ATP-competitive.
  • Chemical Name: 5-(6-Quinoxalinylmethylene)-2,4-thiazolidine-2,4-dione
  • Formula: C12H7N3O2S
  • CAS Number: 648450-29-7
  • Solubility: Soluble to 1mM in DMSO with gentle warming
  • Purity: >98
Description & Specifications

Specifications

AS 605240
>98
50mg
AS 605240
257.27
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit