Learn More
Showa Denko America Inc Asahipak ODP-50 2D DFS Item
Manufacturer:  Showa Denko America Inc F7620009
Reversed phase column, relatively large pore size polyvinyl alcohol base is suitable for the analysis of amino acids, peptides, and proteins, usable in a wide pH range from pH 2 to 13, usable in 100% water and buffer solution, best used for the analysis of basic substances, ODP-50 fulfills USP L67 requirements, Asahipak ODP-50 2D is excellent for LC-MS applications, 2.0 mm ID x 150 mm L
Catalog No. 50-161-3251