Learn More
Showa Denko America Inc Asahipak ODP-50 4D DFS Item
Manufacturer:  Showa Denko America Inc F7620004
Polyvinyl alcohol base material with relatively large pore size is suitable for the analysis of amino acids, peptides, and proteins, usable in a wide pH range from pH 2 to 13, usable in 100% water and buffer solution, best used for the analysis of basic substances, ODP-50 listed under USP L67 requirements
Catalog No. 50-161-3250