Learn More
Showa Denko America Inc Asahipak ODP-50G 2A DFS Item
Manufacturer:  Showa Denko America Inc F6713001
Guard column for Asahipak ODP-50 series 2 mm ID columns, dimensions: 2.0 mm ID x 10 mm L
Catalog No. 50-161-3252