Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ASB-C8Ø, Calbiochem™,

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
216667-49-1
C23H40N2O4S
440.643
AQVJFUZGGMXMED-UHFFFAOYSA-N
C8Ø, 4-n-Octylbenzoylamido-propyl-dimethylammonio Sulfobetaine
3281612
3-[dimethyl-[3-[(4-octylbenzoyl)amino]propyl]azaniumyl]propane-1-sulfonate
CCCCCCCCC1=CC=C(C=C1)C(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-]
216667-49-1
C23H40N2O4S
440.643
AQVJFUZGGMXMED-UHFFFAOYSA-N
C8Ø, 4-n-Octylbenzoylamido-propyl-dimethylammonio Sulfobetaine
3281612
3-[dimethyl-[3-[(4-octylbenzoyl)amino]propyl]azaniumyl]propane-1-sulfonate
CCCCCCCCC1=CC=C(C=C1)C(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-]
Packaging Glass Bottle
Percent Purity ≥95% by HPLC
Color White
Quantity 1g