Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASB12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | ASB12 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17041320
|
Novus Biologicals
NBP17041320UL |
20 μL |
Each for $204.00
|
|
|||||
NBP170413
|
Novus Biologicals
NBP170413 |
100 μL |
Each for $482.50
|
|
|||||
Description
ASB12 Polyclonal specifically detects ASB12 in Human samples. It is validated for Western Blot.Specifications
ASB12 | |
Polyclonal | |
Rabbit | |
ankyrin repeat and SOCS box containing 12, FLJ39577 | |
ASB12 | |
IgG | |
34 kDa |
Western Blot | |
Unconjugated | |
RUO | |
142689 | |
Synthetic peptides corresponding to ASB12(ankyrin repeat and SOCS box-containing 12) The peptide sequence was selected from the C terminal of ASB12. Peptide sequence DDKGIALLLQARATPRSLLSQVRLVVRRALCQAGQPQAINQLDIPPMLIS. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title