Learn More
ASB13 Mouse anti-Human, Clone: OTI3F9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809424
Catalog No. 50-166-9481
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.
Specifications
Specifications
ASB13
Monoclonal
Unconjugated
ASB13
Lyophilized
ASB13
Human recombit protein fragment corresponding to amino acids 76-278 of human ASB13 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3F9
PBS with 8% trehalose and no preservative; pH 7.3
Q8WXK3
ankyrin repeat and SOCS box containing 13
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79754