Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566419
Description
Immunogen sequence: KVKNVDLIEML IEFGGNIYAR DNRGKKPSDY TWSSSAPAKC FEYYEKTPLT LSQLCRVNLR KATGVRGLEK IAKLNI Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.
Specifications
ASB13 | |
Polyclonal | |
Unconjugated | |
ASB13 | |
2210015B19Rik; 6430573K02Rik; ankyrin repeat and SOCS box containing 13; ankyrin repeat and SOCS box protein 13; ankyrin repeat and SOCS box-containing 13; ankyrin repeat and SOCS box-containing protein 13; ankyrin repeat domain-containing SOCS box protein 13; ankyrin repeat domain-containing SOCS box protein Asb-13; ASB13; Asb-13; C85285 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79754 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXK3 | |
ASB13 | |
Recombinant Human ASB13. Recombinant protein control fragment (Product #RP-107091). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction