Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASB17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ASB17 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157066
|
Novus Biologicals
NBP157066 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
ASB17 Polyclonal specifically detects ASB17 in Human samples. It is validated for Western Blot.Specifications
ASB17 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
ankyrin repeat and SOCS box containing 17, ankyrin repeat and SOCS box protein 17, ankyrin repeat and SOCS box-containing 17, Asb-17 | |
ASB17 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q8WXJ9 | |
127247 | |
Synthetic peptides corresponding to ASB17 (ankyrin repeat and SOCS box-containing 17) The peptide sequence was selected from the middle region of ASB17. Peptide sequence SPLSGITPLFYVAQTRQSNIFKILLQYGILEREKNPINIVLTIVLYPSRV. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title