Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ASB4
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095059
View Documents
Novus Biologicals
NBP304635100UL
100 μg
Each for $320.71
Add to cart
 
NB095060
View Documents
Novus Biologicals
NBP30463520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

ASB4 Polyclonal antibody specifically detects ASB4 in Human, Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

ASB4
Unconjugated
Rabbit
Signal Transduction
PBS with 50% glycerol, pH7.3.
51666
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry
Purified
RUO
Human, Mouse
ankyrin repeat and SOCS box containing 4, ankyrin repeat and SOCS box protein 4, ASB-4ankyrin repeat and SOCS box-containing 4, MGC142039, MGC142041
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ASB4 (NP_665879.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit