Learn More
Brady™ Asbestos Warning Pipe Markers, Legend: CAUTION ASBESTOS HAZARD…..TRAINING AND EQUIPMENT
To identifiy asbestos content in pipes. Brady™ Asbestos Warning Pipe Markers identify flow direction and contents at the point of need and are made of durable vinyl suitable for indoor or outdoor environments.
Specifications
Adhesive Type | Acrylic, Permanent, Cold Temperature |
---|---|
Color | Black, Yellow |
Format | Die-cut |
Material | Pressure-sensitive Vinyl (B-946) |
Display Text | CAUTION ASBESTOS HAZARD DO NOT DISTRUB WITHOUT PROPER TRAINING AND EQUIPMENT |
Catalog Number | Mfr. No. | Description | Size (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Size (English) | Price | Quantity & Availability | ||||
19-112-020
|
Brady™
97627 |
Style IHV | 24 x 4 in. (W x H) | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
19-112-021
|
Brady™
97628 |
Style I | 14 x 2.25 in. (W x H) | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
Description
- Includes industrial grade adhesive and peel-to-release coated paper backing
- Identifiy asbestos or asbestos free pipes
Specifications
Acrylic, Permanent, Cold Temperature | |
Die-cut | |
CAUTION ASBESTOS HAZARD DO NOT DISTRUB WITHOUT PROPER TRAINING AND EQUIPMENT |
Black, Yellow | |
Pressure-sensitive Vinyl (B-946) | |
0.004 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.