Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC Polyclonal Antibody, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS34024R

This item is not returnable.
View return policy
Description
ASC Polyclonal antibody specifically detects ASC in Human, Rat samples. It is validated for ELISA, Immunocytochemistry, Western BlotSpecifications
ASC | |
Polyclonal | |
Unconjugated | |
PYCARD | |
9130417A21Rik; Apoptosis-associated speck-like protein containing a CARD; Asc; CARD5; caspase recruitment domain-containing protein 5; hASC; mASC; MGC10332; Pycard; PYD and CARD domain containing; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS1; TMS-1; TNS1 | |
Recombinant human ASC | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q9ULZ3 | |
Rabbit | |
Protein A | |
RUO | |
282817, 29108 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction