Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, Thermo Scientific Chemicals

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage.

Supplier:  Thermo Scientific Chemicals 208090025

 View more versions of this product

Catalog No. AC208090025


Order in Bulk

Request bulk or custom formats

Add to Cart

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
Granular
81133-20-2, 1310-73-2, 7631-86-9
MFCD00132264
Particle size analysis: 20 to 30 mesh
Beige to Brown
20- to 30-mesh
2.5 kg
Glass Bottle
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only