Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, ACROS Organics™

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage.

$149.50 - $1581.49

Specifications

Chemical Name or Material Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
Name Note 20- to 30-mesh
CAS 81133-20-2, 1310-73-2, 7631-86-9
MDL Number MFCD00132264
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC208091000
View Documents
Acros Organics
208091000
100g Glass bottle
Each for $149.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AC208095000
View Documents
Acros Organics
208095000
500g Glass bottle
Each for $505.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AC208090025
View Documents
Acros Organics
208090025
2.5kg Glass bottle
Each for $1,581.49
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage.

Gas Adsorbent
Specifications

Specifications

Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
20- to 30-mesh
81133-20-2, 1310-73-2, 7631-86-9
MFCD00132264
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit