Learn More
Ascarite(II), CO2 absorbent, 8 to 20 mesh, ACROS Organics
Manufacturer: Acros Organics 208081000
Description
Gas AdsorbentSpecifications
Ascarite(II)™, CO2 absorbent, 8 to 20 mesh | |
81133-20-2, 1310-73-2, 7631-86-9 | |
100g | |
Glass bottle | |
Granules |
8- to 20-mesh | |
Brown | |
MFCD00132264 | |
Particle size analysis: 8 to 20 mesh |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.