Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Ascarite(II), CO2 absorbent, 8 to 20 mesh, ACROS Organics™

$154.30 - $2140.00

Specifications

Chemical Name or Material Ascarite(II)™, CO2 absorbent, 8 to 20 mesh
Name Note 8- to 20-mesh
CAS 81133-20-2, 1310-73-2, 7631-86-9
MDL Number MFCD00132264
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC208081000
View Documents
Acros Organics
208081000
100g Each for $154.30
Order in Bulk

Request bulk or custom formats

N/A
AC208085000
View Documents
Acros Organics
208085000
500g Each for $537.00
Order in Bulk

Request bulk or custom formats

N/A
AC208080025
View Documents
Acros Organics
208080025
2.5kg Each for $2,140.00
Order in Bulk

Request bulk or custom formats

N/A
Description

Description

Gas Adsorbent
Specifications

Specifications

Ascarite(II)™, CO2 absorbent, 8 to 20 mesh
8- to 20-mesh
81133-20-2, 1310-73-2, 7631-86-9
MFCD00132264
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit