Learn More
Abnova™ ASCIZ 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00023300T01
Catalog No. 89-935-822
Add to cart
Description
Description
Specifications
Specifications
293T
BC002701.2
pCMV-ASCIZ full-length
100 μL
Human
ASCIZ 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 73.48kDa
Transient overexpression cell lysate was tested with anti-ASCIZ antibody (Mfr. No. H00023300-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot