Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASCL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568843
Description
This target displays homology in the following species: Cow: 92%; Dog: 85%; Guinea Pig: 85%; Horse: 77%; Human: 85%; Mouse: 100%; Rat: 93%.
ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.
Specifications
ASCL3 | |
Polyclonal | |
Unconjugated | |
ASCL3 | |
achaete-scute complex homolog 3; achaete-scute complex homolog 3 (Drosophila); achaete-scute complex homolog-like 3; achaete-scute family bHLH transcription factor 3; achaete-scute homolog 3; Ascl3; ASH-3; ASH3A; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; class A basic helix-loop-helix protein 42; HASH3; ICRFP703B1614Q5.4; ICRFP703N2430Q5.4; mASH3; mASH-3; Sgn1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56787 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9JJR7 | |
ASCL3 | |
synthetic peptide directed towards the middle region of mouse ASCL3. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction