Learn More
Aviva Systems Biology Corp ASCL4 Antibody - N-terminal region (ARP50895_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASCL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASCL4. Alternate names: HASH4, bHLHa44ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60463