Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASCL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557524
Description
Immunogen sequence: METRKPAERL ALPYSLRTAP LGVPGTLPGL PRRDPLRVAL RLDAACWEWA RSGCARGWQY LPVPLDSAFE PAF Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 40%.
ASCL4, a basic helix-loop-helix transcription factor, is essential for the determination of cell fate and the development and differentiation of numerous tissues. It could be a transcriptional regulator involved in skin development.Basic helix-loop-helix transcription factors, such as ASCL4, are essential for the determination of cell fate and the development and differentiation of numerous tissues (Jonsson et al., 2004 [PubMed 15475265]).[supplied by OMIM].
Specifications
ASCL4 | |
Polyclonal | |
Unconjugated | |
ASCL4 | |
achaete-scute complex homolog 4; achaete-scute complex-like 4; achaete-scute family bHLH transcription factor 4; achaete-scute homolog 4; Achaete-scute-like protein 4; ASCL4; ASH-4; BHLHA44; Class A basic helix-loop-helix protein 44; class II bHLH protein ASCL4; HASH4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121549 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6XD76 | |
ASCL4 | |
Recombinant protein corresponding to Human ASCL4. Recombinant protein control fragment (Product #RP-95771). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction