Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascomycin, 95%, Alfa Aesar™

Click to view available options
Quantity:
25mg
100mg
104987-12-4
C43H69NO12
792.02
MFCD06198665
ZDQSOHOQTUFQEM-NURRSENYSA-N
ascomycin,immunomycin,unii-auf4u5nsjk,ascomycin fk520,auf4u5nsjk,chembl8597,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram,ascomycin from streptomyces hygroscopicus var. ascomyceticus,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*,ascomycin fk 520, fr 900520, imm
5282071
CHEBI:29582
CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C
104987-12-4
C43H69NO12
792.02
MFCD06198665
ZDQSOHOQTUFQEM-NURRSENYSA-N
ascomycin,immunomycin,unii-auf4u5nsjk,ascomycin fk520,auf4u5nsjk,chembl8597,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram,ascomycin from streptomyces hygroscopicus var. ascomyceticus,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*,ascomycin fk 520, fr 900520, imm
5282071
CHEBI:29582
CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C

Internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus.

Melting Point 156°C to 160°C
Flash Point >110°C
Quantity 100mg
Packaging Plastic bottle
Boiling Point 868°C
Formula Weight 790
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Ascomycin