Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, Crystalline Powder, 20 to 200 Mesh, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle
50-81-7
C6H8O6
176.124
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
l-ascorbic acid,ascorbic acid,vitamin c,l-ascorbate,ascorbate,ascorbicap,l +-ascorbic acid,cevitamic acid,ascoltin,hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C(C(C1C(=C(C(=O)O1)O)O)O)O
50-81-7
C6H8O6
176.124
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
l-ascorbic acid,ascorbic acid,vitamin c,l-ascorbate,ascorbate,ascorbicap,l +-ascorbic acid,cevitamic acid,ascoltin,hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C(C(C1C(=C(C(=O)O1)O)O)O)O
Quantity 1kg
Mesh Size 20 to 200 Mesh
Specific Gravity 1L = 1.65kg
Packaging Poly Bottle
Formula Weight 176.12
Physical Form Crystalline Powder
Grade FCC/USP
Chemical Name or Material Ascorbic Acid